C12H316CF02E Chemcia Scientific, LLC

2-Hydroxy-3-iodo-4-methoxy-benzaldehyde

  • MDL number MFCD28401496
  • Cas Number : 155560-24-0
  • Molecular Weight : 278.04759

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6192

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6192
Availability details for
Quantity
  • Availability
  • Add to Cart