C12H316CF031 Chemcia Scientific, LLC

5-Amino-4-iodo-2-methyl-phenol

  • MDL number MFCD29067428
  • Cas Number : 388616-53-3
  • Molecular Weight : 249.05231

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Reddish Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6194

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6194
Availability details for
Quantity
  • Availability
  • Add to Cart