C12H316CF032 Chemcia Scientific, LLC

4-Bromo-2-nitro-5-trifluoromethyl-phenol

  • MDL number MFCD29067429
  • Cas Number : 1613719-77-9
  • Molecular Weight : 286.00606

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6195

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6195
Availability details for
Quantity
  • Availability
  • Add to Cart