C12H316CF036 Chemcia Scientific, LLC

8-Bromo-4-oxo-1,4-dihydro-quinoline-2-carboxylic acid

  • MDL number MFCD09937472
  • Cas Number : 1016841-45-4
  • Molecular Weight : 268.06822

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6199

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6199
Availability details for
Quantity
  • Availability
  • Add to Cart