C12H316CF043 Chemcia Scientific, LLC

1-(2H-Indazol-5-yl)-ethanone

  • MDL number MFCD26227386
  • Cas Number : 1001906-63-3
  • Molecular Weight : 160.17691

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6222

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6222
Availability details for
Quantity
  • Availability
  • Add to Cart