C12H316CF047 Chemcia Scientific, LLC

4-Methyl-2H-pyrazolo[3,4-b]pyridin-3-ylamine

  • MDL number MFCD29067436
  • Molecular Weight : 148.16861

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6232

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6232
Availability details for
Quantity
  • Availability
  • Add to Cart