C12H316CF049 Chemcia Scientific, LLC

1-Bromomethyl-4-iodo-2-nitro-benzene

  • MDL number MFCD13178310
  • Cas Number : 375792-98-6
  • Molecular Weight : 341.9318

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6240

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6240
Availability details for
Quantity
  • Availability
  • Add to Cart