C12H316CF063 Chemcia Scientific, LLC

7-Methyl-imidazo[1,2-a]pyridin-6-ylamine

  • MDL number MFCD14547830
  • Cas Number : 1215984-92-1
  • Molecular Weight : 147.18103

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6285

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6285
Availability details for
Quantity
  • Availability
  • Add to Cart