C12H316CF066 Chemcia Scientific, LLC

6-Bromo-5-hydroxy-pyridine-2-carbaldehyde

  • Cas Number : 1273422-87-9
  • Molecular Weight : 202.00828

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 92
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6294

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6294
Availability details for
Quantity
  • Availability
  • Add to Cart