C12H316CF082 Chemcia Scientific, LLC

1-(3-Bromo-2-hydroxy-5-nitro-phenyl)-ethanone

  • Cas Number : 90004-97-0
  • Molecular Weight : 260.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-bromo-2-hydroxy-5-nitrophenyl)ethanone
Inchi Code 1S/C8H6BrNO4/c1-4(11)6-2-5(10(13)14)3-7(9)8(6)12/h2-3,12H,1H3
Inchi Key NUPQONVDLVCQBU-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6345

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6345
Availability details for
Quantity
  • Availability
  • Add to Cart