C12H316CF085 Chemcia Scientific, LLC

6-Methyl-imidazo[1,2-a]pyrazine

  • MDL number MFCD28384400
  • Cas Number : 1346157-11-6
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methylimidazo[1,2-a]pyrazine
Inchi Code 1S/C7H7N3/c1-6-5-10-3-2-8-7(10)4-9-6/h2-5H,1H3
Inchi Key HSKNBNNWHYJGMA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6356

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6356
Availability details for
Quantity
  • Availability
  • Add to Cart