C12H316CF088 Chemcia Scientific, LLC

(3-Bromo-5-nitro-pyridin-4-yl)-ethyl-amine

  • MDL number MFCD11046790
  • Cas Number : 607371-01-7
  • Molecular Weight : 246.06471

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6360

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6360
Availability details for
Quantity
  • Availability
  • Add to Cart