C12H316CF08C Chemcia Scientific, LLC

(4-Methoxy-pyridin-2-yl)-methyl-amine

  • Cas Number : 1104455-24-4
  • Molecular Weight : 138.17055

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6373

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6373
Availability details for
Quantity
  • Availability
  • Add to Cart