C12H316CF08E Chemcia Scientific, LLC

(7-Methoxy-imidazo[1,2-a]pyridin-2-yl)-methanol

  • MDL number MFCD29067469
  • Cas Number : 1781647-84-4
  • Molecular Weight : 178.19225

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Reddish Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6378

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6378
Availability details for
Quantity
  • Availability
  • Add to Cart