C12H316CF08F Chemcia Scientific, LLC

3-Bromo-5-methoxy-imidazo[1,2-a]pyridine

  • Cas Number : 1367950-29-5
  • Molecular Weight : 227.06179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6391

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6391
Availability details for
Quantity
  • Availability
  • Add to Cart