C12H316CF0A7 Chemcia Scientific, LLC

7-Methoxy-imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD12401189
  • Cas Number : 129912-15-8
  • Molecular Weight : 192.17571

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6418

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6418
Availability details for
Quantity
  • Availability
  • Add to Cart