C12H316CF0A9 Chemcia Scientific, LLC

(3-Methyl-1H-pyrazol-4-yl)-methanol

  • MDL number MFCD11100125
  • Cas Number : 51445-36-4
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-methyl-1H-pyrazol-4-yl)methanol
Inchi Code 1S/C5H8N2O/c1-4-5(3-8)2-6-7-4/h2,8H,3H2,1H3,(H,6,7)
Inchi Key HVLNVBVNWISWQC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6430

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6430
Availability details for
Quantity
  • Availability
  • Add to Cart