C12H316CF0AE Chemcia Scientific, LLC

6-Chloro-1H-[1,5]naphthyridin-4-one

  • MDL number MFCD29067476
  • Molecular Weight : 180.59485

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6465

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6465
Availability details for
Quantity
  • Availability
  • Add to Cart