C12H316CF0B2 Chemcia Scientific, LLC

6-Methoxy-4-oxo-1,4-dihydro-[1,5]naphthyridine-3-carboxylic acid ethyl ester

  • MDL number MFCD20922971
  • Cas Number : 724788-63-0
  • Molecular Weight : 248.24044

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6475

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6475
Availability details for
Quantity
  • Availability
  • Add to Cart