C12H316CF0B4 Chemcia Scientific, LLC

4-Oxo-1,4-dihydro-[1,5]naphthyridine-3-carboxylic acid

  • MDL number MFCD20922965
  • Cas Number : 4901-93-3
  • Molecular Weight : 190.15977

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6487

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6487
Availability details for
Quantity
  • Availability
  • Add to Cart