C12H316CF0BB Chemcia Scientific, LLC

1-(5-Methoxy-pyrazin-2-yl)-ethanone

  • MDL number MFCD13190167
  • Cas Number : 320592-61-8
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(5-methoxy-2-pyrazinyl)ethanone
Inchi Code 1S/C7H8N2O2/c1-5(10)6-3-9-7(11-2)4-8-6/h3-4H,1-2H3
Inchi Key SUAQGUZHAQBYPN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6498

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6498
Availability details for
Quantity
  • Availability
  • Add to Cart