C12H316CF0BC Chemcia Scientific, LLC

6-Bromo-3-methoxy-pyrazine-2-carboxylic acid methyl ester

  • MDL number MFCD10697763
  • Cas Number : 259794-06-4
  • Molecular Weight : 247.04944

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6504

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6504
Availability details for
Quantity
  • Availability
  • Add to Cart