C12H316CF0BE Chemcia Scientific, LLC

2-Bromo-4-fluoro-benzothiazole

  • MDL number MFCD03840821
  • Cas Number : 887580-83-8
  • Molecular Weight : 232.07506

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6511

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6511
Availability details for
Quantity
  • Availability
  • Add to Cart