C12H316CF0C7 Chemcia Scientific, LLC

2-Iodo-3-methoxy-benzoic acid

  • MDL number MFCD18904957
  • Cas Number : 50627-31-1
  • Molecular Weight : 278.04759

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6537

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6537
Availability details for
Quantity
  • Availability
  • Add to Cart