C12H316CF0CB Chemcia Scientific, LLC

5-Amino-2-chloro-4-methoxy-benzoic acid

  • Cas Number : 1143622-07-4
  • Molecular Weight : 201.61086

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6545

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6545
Availability details for
Quantity
  • Availability
  • Add to Cart