C12H316CF0CE Chemcia Scientific, LLC

(3,5-Difluoro-phenyl)-acetic acid methyl ester

  • MDL number MFCD12025024
  • Cas Number : 210530-70-4
  • Molecular Weight : 186.15971

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6587

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6587
Availability details for
Quantity
  • Availability
  • Add to Cart