C12H316CF0DF Chemcia Scientific, LLC

3-Amino-5-chloro-pyridine-2-carbonitrile

  • MDL number MFCD07375051
  • Cas Number : 408538-29-4
  • Molecular Weight : 153.57188

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5C
Shipping Temperature Ambient
Country of Origin USA
Product Link http://www.chemcia.com/product_fl02.asp?id=6604

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6604
Availability details for
Quantity
  • Availability
  • Add to Cart