C12H316CF0E5 Chemcia Scientific, LLC

4-Amino-2-chloro-5-iodo-benzoic acid methyl ester

  • MDL number MFCD09260863
  • Cas Number : 256935-85-0
  • Molecular Weight : 311.50789

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6614

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6614
Availability details for
Quantity
  • Availability
  • Add to Cart