C12H316CF0E9 Chemcia Scientific, LLC

4-Amino-2-bromo-benzoic acid

  • MDL number MFCD01569396
  • Cas Number : 2486-52-4
  • Molecular Weight : 216.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-amino-2-bromobenzoic acid
Inchi Code 1S/C7H6BrNO2/c8-6-3-4(9)1-2-5(6)7(10)11/h1-3H,9H2,(H,10,11)
Inchi Key AQBXIOGPHBWBFP-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6620

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6620
Availability details for
Quantity
  • Availability
  • Add to Cart