C12H316CF0ED Chemcia Scientific, LLC

8-Bromo-6-fluoro-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD29764168
  • Molecular Weight : 216.01331

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6624

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6624
Availability details for
Quantity
  • Availability
  • Add to Cart