C12H316CF0F6 Chemcia Scientific, LLC

2-Amino-4-methyl-benzaldehyde

  • MDL number MFCD10696882
  • Cas Number : 59236-38-3
  • Molecular Weight : 135.16703

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6641

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6641
Availability details for
Quantity
  • Availability
  • Add to Cart