C12H316CF0FD Chemcia Scientific, LLC

PYRIDINE-2,3-DIMETHANOL

  • MDL number MFCD01861897
  • Cas Number : 38070-79-0
  • Molecular Weight : 139.15528

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6653

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6653
Availability details for
Quantity
  • Availability
  • Add to Cart