C12H316CF10D Chemcia Scientific, LLC

2-[Bis(tert-Butoxycarbonyl)amino]-6-bromo-benzothiazole

  • MDL number MFCD29764172
  • Molecular Weight : 429.33592

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6670

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6670
Availability details for
Quantity
  • Availability
  • Add to Cart