C12H316CF117 Chemcia Scientific, LLC

5-Amino-4-iodo-2-methyl-benzoic acid methyl ester

  • MDL number MFCD24386972
  • Cas Number : 686342-80-3
  • Molecular Weight : 291.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-amino-4-iodo-2-methylbenzoate
Inchi Code 1S/C9H10INO2/c1-5-3-7(10)8(11)4-6(5)9(12)13-2/h3-4H,11H2,1-2H3
Inchi Key AHPLMUWQBFMNBX-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6687

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6687
Availability details for
Quantity
  • Availability
  • Add to Cart