C12H316CF118 Chemcia Scientific, LLC

5-Amino-4-bromo-2-methyl-benzoic acid methyl ester

  • MDL number MFCD29764173
  • Cas Number : 22378-66-1
  • Molecular Weight : 244.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-amino-4-bromo-2-methylbenzoate
Inchi Code 1S/C9H10BrNO2/c1-5-3-7(10)8(11)4-6(5)9(12)13-2/h3-4H,11H2,1-2H3
Inchi Key DNPYFLRPKSBJDF-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Whit Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6688

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6688
Availability details for
Quantity
  • Availability
  • Add to Cart