C12H316CF119 Chemcia Scientific, LLC

(5-Amino-2-methyl-phenyl)-methanol

  • MDL number MFCD00086622
  • Cas Number : 111437-10-6
  • Molecular Weight : 137.18297

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6689

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6689
Availability details for
Quantity
  • Availability
  • Add to Cart