C12H316CF11A Chemcia Scientific, LLC

(2-Methyl-6-nitro-phenyl)-methanol

  • MDL number MFCD12031847
  • Cas Number : 54915-41-2
  • Molecular Weight : 167.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methyl-6-nitrophenyl)methanol
Inchi Code 1S/C8H9NO3/c1-6-3-2-4-8(9(11)12)7(6)5-10/h2-4,10H,5H2,1H3
Inchi Key LIUYFMADDYMHLM-UHFFFAOYSA-N

Description

Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart