C12H316CF12D Chemcia Scientific, LLC

2-Fluoro-4-nitro-benzamide

  • Cas Number : 350-32-3
  • Molecular Weight : 184.1279

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6733

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6733
Availability details for
Quantity
  • Availability
  • Add to Cart