C12H316CF12E Chemcia Scientific, LLC

(4-Amino-2-fluoro-phenyl)-methanol

  • MDL number MFCD13190556
  • Cas Number : 840501-15-7
  • Molecular Weight : 141.14631

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6736

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6736
Availability details for
Quantity
  • Availability
  • Add to Cart