C12H316CF130 Chemcia Scientific, LLC

(3-Bromo-5-iodo-phenyl)-methanol

  • MDL number MFCD09745849
  • Cas Number : 188813-08-3
  • Molecular Weight : 312.93367

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6740

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6740
Availability details for
Quantity
  • Availability
  • Add to Cart