C12H316CF13D Chemcia Scientific, LLC

7-Chloro-6-iodo-3H-quinazolin-4-one

  • MDL number MFCD28671009
  • Cas Number : 1423187-59-0
  • Molecular Weight : 306.49128

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6829

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6829
Availability details for
Quantity
  • Availability
  • Add to Cart