C12H316CF144 Chemcia Scientific, LLC

4-Bromo-3-formyl-benzonitrile

  • MDL number MFCD11858309
  • Cas Number : 89003-95-2
  • Molecular Weight : 210.03118

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6780

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6780
Availability details for
Quantity
  • Availability
  • Add to Cart