C12H316CF14A Chemcia Scientific, LLC

4-Amino-5-bromo-2-fluoro-benzoic acid methyl ester

  • MDL number MFCD23707475
  • Cas Number : 1427372-46-0
  • Molecular Weight : 248.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-amino-5-bromo-2-fluorobenzoate
Inchi Code 1S/C8H7BrFNO2/c1-13-8(12)4-2-5(9)7(11)3-6(4)10/h2-3H,11H2,1H3
Inchi Key JOCYKKRVIIGZJH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6788

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6788
Availability details for
Quantity
  • Availability
  • Add to Cart