C12H316CF151 Chemcia Scientific, LLC

4-Nitro-thiophene-2-carbonitrile

  • MDL number MFCD03426907
  • Cas Number : 42137-24-6
  • Molecular Weight : 154.14789

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6950

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6950
Availability details for
Quantity
  • Availability
  • Add to Cart