C12H316CF15E Chemcia Scientific, LLC

5-tert-Butyl-2-iodo-phenylamine

  • MDL number MFCD11846052
  • Cas Number : 1160573-98-7
  • Molecular Weight : 275.13418

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6848

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6848
Availability details for
Quantity
  • Availability
  • Add to Cart