C12H316CF160 Chemcia Scientific, LLC

1-Iodo-3-methyl-2-nitro-benzene

  • MDL number MFCD18398068
  • Cas Number : 52414-99-0
  • Molecular Weight : 263.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-iodo-3-methyl-2-nitrobenzene
Inchi Code 1S/C7H6INO2/c1-5-3-2-4-6(8)7(5)9(10)11/h2-4H,1H3
Inchi Key YUHQOZRCAFHTGP-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6860

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6860
Availability details for
Quantity
  • Availability
  • Add to Cart