C12H316CF161 Chemcia Scientific, LLC

N1,3-Dimethylbenzene-1,2-diamine

  • MDL number MFCD19206710
  • Cas Number : 73902-64-4
  • Molecular Weight : 136.19824

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Reddish Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6863

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6863
Availability details for
Quantity
  • Availability
  • Add to Cart