C12H316CF172 Chemcia Scientific, LLC

5-Bromo-6-methyl-2-oxo-1,2-dihydro-pyridine-3-carbonitrile

  • MDL number MFCD20270185
  • Cas Number : 84725-13-3
  • Molecular Weight : 213.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-6-methyl-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
Inchi Code 1S/C7H5BrN2O/c1-4-6(8)2-5(3-9)7(11)10-4/h2H,1H3,(H,10,11)
Inchi Key XSDIDMLVJLNNNV-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6983

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6983
Availability details for
Quantity
  • Availability
  • Add to Cart