C12H316CF176 Chemcia Scientific, LLC

2-Methoxy-6-methyl-pyridine-3-carbaldehyde

  • MDL number MFCD12828119
  • Cas Number : 72918-04-8
  • Molecular Weight : 151.16643

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6989

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6989
Availability details for
Quantity
  • Availability
  • Add to Cart