C12H316CF17C Chemcia Scientific, LLC

3-Nitro-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenylamine

  • MDL number MFCD22493647
  • Cas Number : 928847-00-1
  • Molecular Weight : 264.09129

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7008

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7008
Availability details for
Quantity
  • Availability
  • Add to Cart